ContactUsLinkComponent
Microcon® Centrifugal Filters
Microcon® Centrifugal Filters
Supplier:  Merck
Certificates
undefined
Microcon® Centrifugal Filters
Supplier:  Merck
Specifications
  • Catalog No:
  • MILFMRCPRT010
  • MILFMRCF0R030
  • Filtration area:
  • 0,32 cm²
  • 0,32 cm²
  • Ø×H:
  • 123×450 mm
  • 123×450 mm
  • Sample volume:
  • 500 µl
  • 500 µl
  • Hold-up volume:
  • >10 μl
  • >10 μl