ContactUsLinkComponent
Dishwashing Detergent, ProCare Universal 81 C
Dishwashing Detergent, ProCare Universal 81 C
Catalog # MIEL12456130
Supplier:  MIELE
New Product
Dishwashing Detergent, ProCare Universal 81 C
Catalog # MIEL12456130
Supplier:  MIELE
New Product
Specifications
  • Description:
    Machine descaler, ProCare Universal 81 C
  • Pk:
    250 g
  • Temperature range:
    5...40 °C
Specifications
About this item
Machine descaler, acidic, for removing persistent limescale deposits.
  • Removal of silicate deposits and other acid-soluble impurities
  • Prolongs service life of heater elements
  • Very gentle on materials
  • Very good material compatibility
  • Specially developed for Miele Professional machines
Contains citric acid and does not contain phosphorus compounds.
Certifications: GHS/CLP compliant.