Captive Spray 2 Emitter
Catalog # BRUK1811112
Supplier: BRUKER DALTRONIK
New Product
Specifications
- Description:Captivespray 2 emitter pack
- Pk:1
Specifications
About this item
Our ready-to-use fused silica emitter offers a straightforward plug-and-play setup, ensuring hassle-free connection. Available in two internal diameters, our emitters cover both, high-throughput applications and the specific needs of plasma or single-cell proteomics.
- Already attached ZDV union for easier handling
- For use with the captivespray source, captivespray 2 source or captivespray ultra source
The 10 µm diameter is recommended for applications in the low nano flow range (50 to 450 nl/min).