Order Entry
Switzerland
ContactUsLinkComponent
Plastic Crate with Lid
Plastic Crate with Lid
Catalog # LAMA698
Supplier:  LAMAPLAST
undefined
Plastic Crate with Lid
Catalog # LAMA698
Supplier:  LAMAPLAST

Specifications

  • Capacity:
    70 L
  • Colour:
    Grey
  • Material:
    PP
  • W×D×H:
    600×400×349 mm
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

Crates have two handles on the shorter sides. Crates are light yet strong, providing an economical alternative to metal crates.

  • Polypropylene
  • Colour: Grey
  • For food use