Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Container, PS, with Screw Cap

Container, PS, with Screw Cap

15 ml, 23 mm, 59 mm, Transparent, PS
Catalog # SEMA10728
Supplier:   SEMADENI
Container, PS, with Screw Cap
Catalog # SEMA10728
Supplier:  SEMADENI
Container, PS, with Screw Cap

15 ml, 23 mm, 59 mm, Transparent, PS

Price
SFr. 35.00
Log in for availability
1 * 200 ST
Product Details & Documents

Transparent, with flat bottom and tightly closing screw cap made of polyethylene.

Packaging: Package contains 200 cans and lids individually packed in a bag.

Specifications
  • Ext.Ø:
    23 mm
  • Capacity:
    15 ml
  • Colour:
    Transparent
  • Height:
    59 mm
  • Material:
    PS
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.