Specifications
- Colour:Grey
- Description:Apron model PA30L0, shin-length, neck and waist ties, no seams
- Pk:25
Specifications
About this item
DuPont™ Tychem® 6000 F Apron model PA30L0. Shin-length apron with neck and waist ties. Available in grey and in one size.
Specially designed for use with Tychem® apparel, Tychem® accessories can help offer enhanced protection for body parts that are more exposed to hazardous substances.
Tychem® 6000 F garments and accessories are made from a lightweight and durable fabric consisting of a proprietary barrier film laminated to a heavy duty Tyvek® substrate. They offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
Tychem® 6000 F is used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.