ContactUsLinkComponent
Savant™ SpeedVac™ SPD120P2 Vacuum Concentrator Kit
  THSASPD120P2-230
undefined
Savant™ SpeedVac™ SPD120P2 Vacuum Concentrator Kit
  THSASPD120P2-230
  • Description:
    SpeedVac™ SPD120P2 vacuum concentrator kit, ambient, 35...80˚C in 5 °C increments, 230 V, 50 Hz
  • W×D×H:
    33×46×36 cm
  • Weight:
    26 kg
  • Pk:
    1
The Thermo Scientific™ Savant™ SpeedVac™ SPD120 vacuum concentrator can be used with a wide variety of solvents and offers resistance to methanol, acetonitrile with 0,1% TFA, and other aggressive solvents used in DNA and biological samples. With a medium-capacity, modular design, the SPD120 vacuum concentrator can also be used for applications where freeze-drying or lyophilisation is needed.
  • Basic molecular biology applications with increased microplate capacity
  • Simple display and programming
  • Six programs available
  • Enhanced evaporation model, organic compatible
  • Radiant glass cover to accelerate evaporation and reduce drying time
  • Vacuum end point mode terminates run when preset vacuum level is reached
  • Digital display of vacuum, temperature and time section
  • Latched lid and lid lock indicator ensuring user safety during operation
Solvent and solvent combinations: Acetonitrile, Ethanol, Isopropanol, Methanol, Water, Acetonitrile and water, Ethanol and water, Methanol and water, and Ammonium hydroxide.
 : SPD120P2 includes acid-resistant SpeedVac™ vacuum concentrator, ultra-low temperature refrigerated vapour trap (–105 °C ), vacuum pump, digital vacuum gauge, chemical trap cartridge, cartridge for vapour trap, glass tubes rotor, RH40-11 (40×1.5 to 2.0 ml), Glass condensation flask, tubing and fitting kit, CryoCool™ heat transfer fluid (1 L).