Order Entry
Switzerland
ContactUsLinkComponent
Zinc acetate 0.25 mol/l Reag. Ph. Eur. 1102301, buffered with ammonium acetate pH 6.4
Environment
Zinc acetate 0.25 mol/l Reag. Ph. Eur. 1102301, buffered with ammonium acetate pH 6.4
Catalog # 87967.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Environment
Zinc acetate 0.25 mol/l Reag. Ph. Eur. 1102301, buffered with ammonium acetate pH 6.4
Catalog # 87967.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :183.48 g/mol
Storage Temperature:Ambient
EINECS:209-170-2
UN:3082
ADR:9,III
 :GHS09
CAS Number:557-34-6
Formula:C₄H₆O₄Zn
MDL Number:MFCD00012454