Order Entry
Switzerland
ContactUsLinkComponent
Sodium hydroxide 0.005 mol/l (0.005 N) in methanol 50% Reag. Ph. Eur. 1081405
Flammable
Toxic
Health
Sodium hydroxide 0.005 mol/l (0.005 N) in methanol 50% Reag. Ph. Eur. 1081405
Catalog # 87937.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Toxic
Health
Sodium hydroxide 0.005 mol/l (0.005 N) in methanol 50% Reag. Ph. Eur. 1081405
Catalog # 87937.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
UN:3286
ADR:3 (6.1, 8),II
 :GHS02
 :GHS06
 :GHS08
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548