Order Entry
Switzerland
ContactUsLinkComponent
Titanium(III) chloride 15% in hydrochloric acid 10% Reag. Ph. Eur. 1091201
Corrosive
Irritant
Titanium(III) chloride 15% in hydrochloric acid 10% Reag. Ph. Eur. 1091201
Catalog # 87708.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
Titanium(III) chloride 15% in hydrochloric acid 10% Reag. Ph. Eur. 1091201
Catalog # 87708.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :154.24 g/mol
EINECS:231-728-9
UN:1789
ADR:8,II
 :GHS05
 :GHS07
CAS Number:7705-07-9
Formula:Cl₃Ti
MDL Number:MFCD00011266