Vaseline, Solidification point: 50…55 °C, white, meets analytical specification of Ph.Eur.
Supplier: Roth Carl
Certificates
About this item
Synonyms: Petrolatum, Petroleum Jelly
CAS Number:8009-03-8
:
Melting Pt:55 °C
Density:0.94 g/cm³ (25 °C)
EINECS:295-456-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:white to off-white
- Identity:complies
- Solidification point:50-55 °C
- Viscosity (100 °C):7,5-10 mm2/s
- Aromatic polycyclic hydrocarbons:complies
- Sulphated ash:≤0,05 %
- Consistency:165-185 1/10 mm
- Acid or alkaline react. substances:complies
Specification Test Results
Frequently Bought Together