
Celite® 535, filter aid
Anbieter: MACHEREY-NAGEL
Zertifikate
Über diesen Artikel
CAS-Nummer:68855-54-9
:
Formel:SiO₂
MDL-Nummer:MFCD00132803
:60.08 g/mol
:GHS08
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Anbieter: VWR Chemicals
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
VWR®, Nitrile Gloves
Anbieter: VWR Collection
VWR®, Slide Boxes
Anbieter: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
VWR®, Short Thread Vials ND9
Anbieter: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Anbieter: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Anbieter: VWR Collection
VWR®, Pasteur Pipettes
Anbieter: VWR Collection
Xylene (mixture of isomers) ≥98%, TECHNICAL
Anbieter: VWR Chemicals
VWR®, Screw Neck Vials ND13
Anbieter: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals