Order Entry
Switzerland
ContactUsLinkComponent
Conductivity standard 12880 µS/cm (25 °C; 0.1 mol/L; KCl) for conductivity measurement
Conductivity standard 12880 µS/cm (25 °C; 0.1 mol/L; KCl) for conductivity measurement
Catalog # KNICCS-C12880K/500
Supplier:  KNICK
CAS Number:  
Conductivity standard 12880 µS/cm (25 °C; 0.1 mol/L; KCl) for conductivity measurement
Catalog # KNICCS-C12880K/500
Supplier:  KNICK
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    Conductivity standard, (0,1 mol/l KCl), ready-to-use
  • Seq:
    40
  • Conductivity:
    12880 µS/cm

Specifications

About this item