Ammonia solution 7 N in methanol, Fluka™
Glass bottle, 1 L
Catalog # 499145-1L
Supplier: Honeywell Chemicals
Ammonia solution 7 N in methanol, Fluka™
Catalog # 499145-1L
Supplier: Honeywell Chemicals
CAS Number: 1336-21-6
Jump to:
Product Details & Documents
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00011418
- Molecular Weight:35.05 g/mol
- Density:0.779 g/cm3 (25 °C)
- Flash Pt:14 °C
- Storage Temperature:Ambient
- EINECS:215-647-6
- UN:3286
- ADR:3,II
Specifications
Specification Test Results
- Assay:6.5 - 7.5 mol/l
- Water (Karl Fischer):Max. 0.1 %
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



