Order Entry
Switzerland
ContactUsLinkComponent
Pyridoxine hydrochloride
Catalog # EHERC16652000
Supplier:  EHRENSTORFER
CAS Number:  
Pyridoxine hydrochloride
Catalog # EHERC16652000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :205.64 g/mol
Merck Index:13,08072
Melting Pt:214…215 °C
Storage Temperature:Ambient
EINECS:200-386-2
CAS Number:58-56-0
Formula:C₈H₁₂ClNO₃
MDL Number:MFCD00012807