ContactUsLinkComponent
Thermo Scientific™ Teleshake Magnetic Shaking System
Thermo Scientific™ Teleshake Magnetic Shaking System
Catalog # HERA50134882
Supplier:  Thermo Scientific
undefined
Thermo Scientific™ Teleshake Magnetic Shaking System
Catalog # HERA50134882
Supplier:  Thermo Scientific
Specifications
  • Description:
    Teleshake magnetic shaking system without controller
  • Pk:
    1
Specifications
About this item
Conduct powerful shaking applications from 100 to 2000rpm with the Thermo Scientific™ Teleshake magnetic shaking system, a remote-controlled, wear-free magnetic shaking system for integrated shaking applications; suited for 96 and 384 well microtiter plates.
  • Easy integration in robotic systems and platform due to small footprint
  • Simultaneous control of up to 14 shakers via RS232 interface
  • Powerful shaking movements from 100 to 2000 min⁻¹ (rpm), with an amplitude of 2 mm
  • Rapid switching between circular and linear agitation
  • Minimum of moved masses, therefore little transference of vibrations onto the installation area
  • Integrated threads and drilled holes for assembling on robot workbenches