Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Test Tubes

Test Tubes

8 ml, 61 mm, 16,2 mm, 15-425
Catalog # METB30210021
Supplier:   Mettler - Toledo
Test Tubes
Catalog # METB30210021
Supplier:  Mettler - Toledo
Test Tubes

8 ml, 61 mm, 16,2 mm, 15-425

Price
Inquire For Price
Log in for availability
1 * 100 ST
Product Details & Documents

Test tube with 15-425 thread.

  • Made of borosilicate glass
Specifications
  • Ø:
    16,2 mm
  • Thread:
    15-425
  • Capacity:
    8 ml
  • Length:
    61 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.