Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
Switzerland
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
ContactUsLinkComponent
Home
Profile solution
Profile solution detergent, for cleaning, chlorinated
Profile solution detergent, for cleaning, chlorinated
Profile solution detergent, for cleaning, chlorinated
Katalognr. DIVY101107114
Supplier:
Diversey
New Product
CAS Number:
Expand image
Profile solution detergent, for cleaning, chlorinated
Katalognr. DIVY101107114
Supplier:
Diversey
New Product
CAS Number:
Specifications
Pack type:
Plastic can
Pk:
20 L
Seq:
5302
Specifications
About this item
Product Details
:
Product Details