Anti-Androgen Receptor (AR-V7 specific) Rabbit Monoclonal Antibody [clone: DHH-1]
Catalog # BSBTM00542-3
Supplier: Boster Bio
New Product
Specifications
- Antigen-Name:Androgen Receptor (AR-V7 specific)
- Klonalität:Monoklonal
- Klon:DHH-1
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Form:Liquid
- Antigen-Synonyme:TFM|nuclear receptor subfamily 3 group C member 4|HYSP1|AR androgen receptor androgen receptor|Androgen receptor|dihydrotestosterone receptor|SBMA|SMAX1|AR|Nuclear receptor subfamily 3 group C member 4|NR3C4|AR AIS8|HUMARA|KD|Dihydrotestosterone receptor|DHTR
- Lagerungspuffer:Rabbit IgG in phosphate buffered saline, pH 7,4, 150 mM NaCl, 0,02% sodium azide and 50% glycerol, 0,4 - 0,5 mg/ml BSA.
- Molecular weight:99188
- Lagertemperatur:Store at –20 °C for one year. For short term storage and frequent use, store at 4 °C for up to one month. Avoid repeated freeze-thaw cycles.
- Immunogen:A synthesised peptide derived from human Androgen receptor (AR-V7 specific).
- VE:100 µl
Specifications
About this item
Type: Primary
Antigen: Androgen Receptor (AR-V7 specific)
Clonality: Monoclonal
Clone: DHH-1
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human