ContactUsLinkComponent
Workstation Spilltray and Drying Rack
Workstation Spilltray and Drying Rack
  STUA67035-01
New Product
Workstation Spilltray and Drying Rack
  STUA67035-01
New Product
  • Description:
    Workstation spilltray and drying rack
  • Pk:
    1
  • W×H:
    330,2×15,9 mm
Reduce drying time and contain spilled liquids with this chemical resistant containment tray. Holds one ¹/₄ L of spilled liquid. Easy to clean simply lift off the white grid and empty.
  • Chemical resistant
  • Easy to clean
  • Dishwasher safe
  • Material: Polyethylene(PE)
  • Length: 419 mm
 : One year warranty.