Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Phenylthiohydantoinalanine
Phenylthiohydantoinalanine ≥98.0% (by HPLC)
Phenylthiohydantoinalanine ≥98.0% (by HPLC)
TCIAP0374.1
:
TCI
:
Phenylthiohydantoinalanine ≥98.0% (by HPLC)
TCIAP0374.1
:
TCI
:
Pk:
1 g
Seq:
3047
Purity/Analysis Method:
>98.0% (HPLC)
:
:
206.26 g/mol
:
187 °C
:
4333-19-1
:
C₁₀H₁₀N₂OS
:
MFCD06858229