Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Silicate (Silicic Acid) Test Refill pack, Supelco® MQuant®, Merck

Silicate (Silicic Acid) Test Refill pack, Supelco® MQuant®, Merck

Refill pack for the MQuant® test kit 1.18765, Colorimetric, for use with MCOLORTEST®, 100 Tests
Catalog # 1.08045.0001
Supplier:   Merck
Silicate (Silicic Acid) Test Refill pack, Supelco® MQuant®, Merck
Catalog # 1.08045.0001
Supplier:  Merck
CAS Number:  
Silicate (Silicic Acid) Test Refill pack, Supelco® MQuant®, Merck
Silicate (Silicic Acid) Test Refill pack, Supelco® MQuant®, Merck

Refill pack for the MQuant® test kit 1.18765, Colorimetric, for use with MCOLORTEST®, 100 Tests

CAS Number:  
Price
73.40 SFr.
Log in for availability
1 * 100 Tests
Product Details & Documents

This refill pack contains all the reagents for the MQuant® test kit 1.18765 for the colorimetric evaluation of silicate. When the reagents of the MQuant® test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MQuant® test kits, making it a particularly economical system.

  • Colorimetric
  • Microbialanalysis

The test can be used for the determination of silicate e.g. in groundwater and surface water, drinking water and industrial water.

Documents

Specifications

Specifications

  • Chemical method:
    Colorimetric
  • Pk:
    100 Tests
  • Description:
    Refill pack for the MQuant® test kit 1.18765
  • For unit:
    for use with MCOLORTEST®
  • Cat. No.:
    1.08045.0001
  • M-Clarity quality level:
    MQ100
  • Storage temperature:
    15…25 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.