ContactUsLinkComponent
AmberChrom™ chloride form 1x2 100-200 mesh
AmberChrom™ chloride form 1x2 100-200 mesh
New Product
Certificates
AmberChrom™ chloride form 1x2 100-200 mesh
New Product
Specification Test Results
  • Appearance:
    white to yellow beads
  • Total exchange capacity:
    ≥0.6 eq/l (Cl form)
  • Water retention capacity:
    65 to 75 % (Cl form)
  • Particle size:
    100 to 200 mesh