ContactUsLinkComponent
AmberChrom™ hydrogen form 50WX2 200-400 mesh
Corrosive
AmberChrom™ hydrogen form 50WX2 200-400 mesh
New Product
Certificates
Corrosive
AmberChrom™ hydrogen form 50WX2 200-400 mesh
New Product
Specification Test Results
  • Appearance:
    white to yellow to orange beads
  • Total exchange capacity:
    ≥0.6 eq/l (H form)
  • Water retention capacity:
    74 to 82 % (H form)
  • Particle size:
    200 to 400 mesh