AmberChrom™ chloride form 1x8 100-200 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69011-19-4
:MFCD00132718
Formula:(C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
Storage Temperature:Ambient
Specification Test Results
- Appearance:white to yellow beads
- Total exchange capacity:≥1.2 eq/l (Cl form)
- Water retention capacity:39 to 45 % (Cl form)
- Particle size:100 to 200 mesh
Specification Test Results
Frequently Bought Together