ContactUsLinkComponent
AmberChrom™ chloride form 1x8 50-100 mesh
AmberChrom™ chloride form 1x8 50-100 mesh
New Product
Certificates
AmberChrom™ chloride form 1x8 50-100 mesh
New Product
Specification Test Results
  • Appearance:
    white to yellow to tan beads. Material can contain black specks per manufacturer. Does not impact use.
  • Total exchange capacity:
    ≥1.2 eq/l (Cl form)
  • Water retention capacity:
    43 to 48 % (Cl form)
  • Particle size:
    50 to 100 mesh