AmberChrom™ hydrogen form 50WX8 100-200 mesh
New Product
:69011-22-9
:
:MFCD00132726
:GHS05
:Ambient
- Appearance:beige to green-beige to light brown beads
- Total exchange capacity:≥1.7 eq/l (H form)
- Water retention capacity:50 to 58 % (H form)
- Particle size:100 to 200 mesh
Frequently Bought Together