
AmberChrom™ hydrogen form 50WX2 100-200 mesh
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:69011-22-9
:
MDL Number:MFCD00132722
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:white to yellow to orange beads
- Total exchange capacity:≥0.6 eq/l (H form)
- Water retention capacity:74 to 82 % (H form)
- Particle size:100 to 200 mesh
Specification Test Results
Frequently Bought Together