
Nutlin-3 98%
Catalog # 469201000.
Supplier: Thermo Fisher Scientific
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:white to light yellow crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Loss on drying:≤5 %
Specification Test Results
About this item
:
:581.49 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:548472-68-0
Formula:C₃₀H₃₀Cl₂N₄O₄
MDL Number:MFCD07784509