
Methyl-3-hydroxyisoxazole-5-carboxylate 97+%
Catalog # 263080050.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:white to light yellow crystalline powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥97.0 %
- Melting point:158°C to 163°C
Specification Test Results
About this item
:
:143.1000 g/mol
Boiling Pt:261.1°C
:GHS07
Storage Temperature:Ambient
CAS Number:10068-07-2
Formula:C₅H₅NO₄
MDL Number:MFCD00062870