Order Entry
Switzerland
ContactUsLinkComponent
NGL-IMPACT® CH1 Affinity Ligand ELISA Kit
NGL-IMPACT® CH1 Affinity Ligand ELISA Kit
Catalog # SPEC9-EL-0040
New Product
CAS Number:  
NGL-IMPACT® CH1 Affinity Ligand ELISA Kit
Catalog # SPEC9-EL-0040
New Product
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    CH1 affinity ligand ELISA kit
  • Sensitivity:
    0,1 ng/ml

Specifications

About this item

The NGL-IMPACT® CH1 Affinity Ligand ELISA Kit provides accurate and precise quantitation for downstream processes that utilise a Purolite Praesto Jetted 70 CH1 capture step.

  • Exact matched reference standard (NGL-IMPACT® CH1 ligand)
  • Recovery: 80 to 120%
  • Accuracy and precision: inter-assay ±15%, intra-assay ±15%

Kit contents: Anti-CH1 antibody coated 96 well microtiter plate, Reagent A: sample diluent (5X), Reagent B: NGL-IMPACT® CH1 Ligand standard solution, Reagent C: rabbit anti-CH1: biotin probe, Reagent D: streptavidin: peroxidase conjugate, Reagent E: TMB peroxidase substrate, PBS packs (quantity 2)