Order Entry
Switzerland
ContactUsLinkComponent
Weighing Platform, PBA639-B30
Weighing Platform, PBA639-B30
Catalog # METB30664649
Supplier:  Mettler - Toledo
New Product
Weighing Platform, PBA639-B30
Catalog # METB30664649
Supplier:  Mettler - Toledo
New Product

Specifications

  • Model:
    PBA639-B30
  • Pan/Platform Size:
    500L×400W×126H mm
  • Description:
    Weighing platform, stainless steel, analog interface with 2,5 m cable
  • Pk:
    1
  • Readability:
    1 g
  • Weighing cap.:
    15/30 kg
  • Repeatability:
    0,28 g

Specifications

About this item

The PBA639 stainless-steel weighing platform strictly adheres to hygienic design guidelines, facilitates quick and easy cleaning and meets higher accuracy standards to optimise processes and reduce costs in regulated hygienic environments.

  • Cleaning and compliance made easy
  • No cavities or crevices
  • Hazardous area approved
  • Fully protected load cell

The elimination of open threads, cavities and dead spots helps to prevent contamination by giving bacteria nowhere to hide. This stainless-steel, hermetically sealed load cell withstands harsh and wet environments with IP68 and IP69k protection.