Order Entry
Switzerland
ContactUsLinkComponent
Zinc, AAS Standard Solution
Zinc, AAS Standard Solution
Catalog # ACSDA069.2NP.L1
Supplier:  CPACHEM
New Product
CAS Number:  
Zinc, AAS Standard Solution
Catalog # ACSDA069.2NP.L1
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Storage conditions:
    15...25 °C
  • Shelf life:
    36 Months
  • Element:
    Zinc
  • Symbol:
    Zn

Specifications

About this item

Certifications: ISO 17034, ISO 17025, ISO 9001