Order Entry
Switzerland
ContactUsLinkComponent
Arsenic Standard Solution, 1000 mg/L in Diluted HNO₃ for ICP/MS
Arsenic Standard Solution, 1000 mg/L in Diluted HNO₃ for ICP/MS
N° de catalogue ACSDM203.2NP.L1
Fournisseur:  CPACHEM
New Product
Numéro CAS:  
Arsenic Standard Solution, 1000 mg/L in Diluted HNO₃ for ICP/MS
N° de catalogue ACSDM203.2NP.L1
Fournisseur:  CPACHEM
New Product
Numéro CAS:  

Spécifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Description:
    Arsenic
  • Symbol:
    As

Spécifications

À propos de cet article

Certifications: ISO 17034, 17025, 9001 certified.