Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Nalidixic acid

Nalidixic acid

100 mg
Catalog # ACSDSB39240.100MG
Supplier:   CPACHEM
Nalidixic acid
Catalog # ACSDSB39240.100MG
Supplier:  CPACHEM
CAS Number:  389-08-2
Nalidixic acid

100 mg

CAS Number:  389-08-2
Price
139.00 SFr.
Log in for availability
1 * 100 mg
Product Details & Documents

Certifications: ISO 17034, 17025, 9001 certified.

  • CAS Number:389-08-2
  • Formula:C₁₂H₁₂N₂O₃
  • MDL Number:MFCD00006884
  • Molecular Weight:232.24 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pk:
    100 mg

Specifications