Order Entry
Switzerland
ContactUsLinkComponent
 
Nitric acid diluted, Test Solution (TS)
Catalog # ACSDCH8002091.L1
Supplier:  CPACHEM
New Product
CAS Number:  
Nitric acid diluted, Test Solution (TS)
Catalog # ACSDCH8002091.L1
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :63.01 g/mol
Storage Temperature:Room Temperature
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349