Order Entry
Switzerland
ContactUsLinkComponent
 
Irritant
for cellulose dissolution
Catalog # 31115.267
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
for cellulose dissolution
Catalog # 31115.267
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :165.68 g/mol
EINECS:241-508-4
UN:3266
ADR:8,III
 :GHS07
 :17500-49-1
Formula:CuH₁₄N₄O₂