Order Entry
Switzerland
ContactUsLinkComponent
Sodium picrate 13 g/l in aqueous solution Reag. Ph. Eur. 1065802
Sodium picrate 13 g/l in aqueous solution Reag. Ph. Eur. 1065802
Catalog # 87715.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium picrate 13 g/l in aqueous solution Reag. Ph. Eur. 1065802
Catalog # 87715.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :251.09 g/mol
EINECS:222-038-9
CAS Number:3324-58-1
Formula:C₆H₂N₃NaO₇