SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Hydrochloric acid
Hydrochloric acid 2 mol/l (2 N) Reag. Ph. Eur. 1043503
Hydrochloric acid 2 mol/l (2 N) Reag. Ph. Eur. 1043503
85848.290
:
VWR Chemicals
:
Hydrochloric acid 2 mol/l (2 N) Reag. Ph. Eur. 1043503
85848.290
:
VWR Chemicals
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
:
:
1.031 g/cm³ (20 °C)
:
Ambient
:
231-595-7
:
8,III
:
36.46 g/mol
:
1789
:
GHS05
:
7647-01-0
:
HCl
:
MFCD00011324