ContactUsLinkComponent
VWR® Conductivity standard, CRM, 5000 μS/cm
VWR® Conductivity standard, CRM, 5000 μS/cm
Catalog # 85645.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Conductivity standard, CRM, 5000 μS/cm
Catalog # 85645.260
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    KCl 0,0375 mol/l: 5000 µS/cm
  • Seq:
    26
Specifications
About this item
Directly traceable to NIST, tested and certified to ISO17025 and ISO 17034