Order Entry
Switzerland
ContactUsLinkComponent
Fehling's reagent (kit, contains solution I and II) Reag. Ph. Eur. 1023300 for qualitative determination of reducing sugars
Fehling's reagent (kit, contains solution I and II) Reag. Ph. Eur. 1023300 for qualitative determination of reducing sugars
Catalog # 87814.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Fehling's reagent (kit, contains solution I and II) Reag. Ph. Eur. 1023300 for qualitative determination of reducing sugars
Catalog # 87814.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 KIT
  • Seq:
    2735
  • Pk Info:
    2x500 ml

Specifications

About this item

 :
UN:3316
ADR:9,II