Order Entry
Switzerland
ContactUsLinkComponent
Tin(II) chloride 30% in hydrochloric acid 20% Reag. Ph. Eur. 1085001
Corrosive
Tin(II) chloride 30% in hydrochloric acid 20% Reag. Ph. Eur. 1085001
Catalog # 87941.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Tin(II) chloride 30% in hydrochloric acid 20% Reag. Ph. Eur. 1085001
Catalog # 87941.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :189.62 g/mol
EINECS:231-868-0
UN:1789
ADR:8,II
 :GHS05
CAS Number:7772-99-8
Formula:SnCl₂
MDL Number:MFCD00011241