Order Entry
Switzerland
ContactUsLinkComponent
Savana® Semiconductor Lab Disc Disposable Filter
Savana® Semiconductor Lab Disc Disposable Filter
Catalog # ENTESVL13D500QQA25
Supplier:  ENTEGRIS
New Product
Savana® Semiconductor Lab Disc Disposable Filter
Catalog # ENTESVL13D500QQA25
Supplier:  ENTEGRIS
New Product

Specifications

  • Porosity:
    5,0 µm
  • Description:
    Dispoable filter, SL 13, PP
  • Pk:
    1

Specifications

About this item

Savana® Lab Disc disposable filters have been specially designed for simple, quick and efficient filtration of liquids used in laboratory, pilot and small-scale applications.

  • Inlet fitting: BD Luer-lok female
  • Outlet fitting: BD Luer-lok female
  • Size: 19 mm

Eight different media options with fitting options can be placed in an all-polypropylene or polyethylene shell construction for excellent chemical compatibility. The small, compact design of the filter capsule also reduces hold-up volume and exposure to hazardous chemicals. No adhesives, binders or surfactants are used in the manufacturing process. The unit is thermally sealed to ensure integrity.

Packaging: Pack of 25.