Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Albert color kit
Albert color kit stain for bacteriology
Albert color kit stain for bacteriology
Albert color kit stain for bacteriology
Catalog # LIOF80294
Supplier:
LIOFILCHEM
New Product
CAS Number:
Expand image
Albert color kit stain for bacteriology
Catalog # LIOF80294
Supplier:
LIOFILCHEM
New Product
CAS Number:
Specifications
Pk:
1 KIT
Seq:
2593
Pk Info:
2×250 ml
Specifications
About this item
Product Details
:
:
1
Product Details