ContactUsLinkComponent
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 99+% for molecular biology DNase-, RNase-, protease-free
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 99+% for molecular biology DNase-, RNase-, protease-free
Certificates
undefined
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 99+% for molecular biology DNase-, RNase-, protease-free
Specification Test Results
  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.2 %
  • TLC:
    ≥99.0 %
  • Specific optical rotation:
    -61° to -63° (20°C, 589 nm) (c=1, H2O/DMF 1/1)
  • Dnase,Rnase,Protease activity:
    none detected