ContactUsLinkComponent
AlphaTec® 87-118 Natural Rubber Latex Gloves
AlphaTec® 87-118 Natural Rubber Latex Gloves
Supplier:  Ansell
New Product
Certificates
undefined
AlphaTec® 87-118 Natural Rubber Latex Gloves
Supplier:  Ansell
New Product
Specifications
  • Catalog No:
  • ANSE87118/6.5
  • ANSE87118/7.5
  • ANSE87118/9.5
  • ANSE87118/8.5
  • ANSE87118/10.5
  • Latex / latex-free:
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Liner material:
  • Cotton
  • Cotton
  • Cotton
  • Cotton
  • Cotton
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Palm thickness:
  • 0,8 mm
  • 0,8 mm
  • 0,8 mm
  • 0,8 mm
  • 0,8 mm