Order Entry
Switzerland
ContactUsLinkComponent
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
Irritant
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
Catalog # MOLE71674020-25G
Supplier:  Molekula
CAS Number:  
undefined
Irritant
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
Catalog # MOLE71674020-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :191.7 g/mol
Melting Pt:112…113 °C
EINECS:247-361-2
 :GHS07
Storage Temperature:Freezer
CAS Number:25952-53-8
Formula:C₈H₁₇N₃·HCl
MDL Number:MFCD00012503