Order Entry
Switzerland
ContactUsLinkComponent
Pyridoxine hydrochloride
Pyridoxine hydrochloride
Catalog # PROOTRCP991735-1G
New Product
CAS Number:  
Pyridoxine hydrochloride
Catalog # PROOTRCP991735-1G
New Product
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.64 g/mol
Storage Temperature:Freezer
CAS Number:58-56-0
Formula:C₈H₁₂ClNO₃
MDL Number:MFCD00012807