Tetradecyltrimethylammonium bromide ≥99% (by titration), Calbiochem®, Sigma-Aldrich®
Catalog # 324411-500
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mg
- Seq:2725
- Environmentally Preferable:
Specifications
About this item
Synonyms: MiTMAB, (1-Tetradecyl)trimethylammonium bromide, N,N,N-Trimethyltetradecan-1-aminium bromide, Tetradonium bromide, Tetradecyl trimethyl ammonium Bromide, Myristyltrimethylammonium bromide, Dynamin Inhibitor II, TTAB
:
:336.394 g/mol
:M-Clarity™ quality level = MQ100
:GHS08
:GHS05
:GHS07
:GHS09
Storage Temperature:Refrigerator
CAS Number:1119-97-7
:8044-71-1
Formula:C₁₇H₃₈BrN
MDL Number:MFCD00011770