ContactUsLinkComponent
Print-on Hook Labels BradyGrip® with Velcro® Hook for BradyJet™ J7300 Printer
Print-on Hook Labels BradyGrip® with Velcro® Hook for BradyJet™ J7300 Printer
Catalog # BRDY176590
Supplier:  Brady
undefined
Print-on Hook Labels BradyGrip® with Velcro® Hook for BradyJet™ J7300 Printer
Catalog # BRDY176590
Supplier:  Brady
Specifications
  • Type:
    B-2414
  • Colour:
    White
  • Dimensions:
    25,40W mm × 22,86L m
  • Material:
    Polyester
  • Model:
    JC-1000-2414
  • Pk:
    1
Specifications
About this item
BradyGrip® Print-on Hook labels combine the power of on-demand colour labelling and the flexibility of VELCRO® material for unmatched identification of wires and cables.
  • Keep wires and cables clearly identified to breeze through installations, troubleshooting, and repairs
  • Use colour to add strong visual cues when locating wire and cable bundles
  • Removable and repositionable
  • Inkjet receptive polyester material
  • Designed for indoor use
Easily apply, reposition and remove this exclusive print-and-place material created from a Brady and VELCRO® brand partnership. Simply print a unique label, then apply to a VELCRO® Brand ONE-WRAP® roll. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 colour label printer. Ink cartridges are required to print this material; use part numbers P-CMY-1 and P-K-1. VELCRO® Brand ONE-WRAP® product not included with purchase.